Learn More
Medchemexpress LLC GNE-235 | 3036574-72-5 | C13H16N4O | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY150289
Encompass_Preferred
GNE-235 is a compound selective for the second bromodomain of PBRM1, with a KD of 0.28 ± 0.02 μM. It can be used for the evaluation of the cellular function of PBRM1.
  • Selective for the second bromodomain of PBRM1
  • KD of 0.28 ± 0.02 μM
  • Useful for evaluating the cellular function of PBRM1
  • Purity: 98.45%
Catalog No. 50-003-74299