Learn More
Medchemexpress LLC GNE-477 | 1032754-81-6 | 98.8% | 504.63 | 10 MG

Supplier: Medchemexpress LLC HY1104210MM/1ML
GNE-477 is a potent and efficacious dual PI3K (IC50=4 nM)/mTOR(Ki=21 nM) inhibitor.
- Potent and efficacious dual PI3K/mTOR inhibitor
- Available as a solid or a solution in DMSO
- Powder can be stored at -20°C for 3 years or 4°C for 2 years
- Solution in solvent can be stored at -80°C for 2 years or -20°C for 1 year
- Soluble in DMSO at 16.67 mg/mL (33.03 mM), requiring ultrasonic treatment
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.