Learn More
Selleck Chemical LLC GNE-495 E4722-25MG
SDP
Supplier:  Selleck Chemical LLC E472225MG
Encompass_Preferred
GNE-495 is a potent and selective inhibitor of MAP4K4 with an IC50 value of 3 7 nM It demonstrates excellent potency and good Pharmacokinetics in in vivo efficacy in a retinal angiogenesis model
Catalog No. 50-000-39535