Learn More
Selleck Chemical LLC GNE-781
SDP
Supplier:  Selleck Chemical LLC S8665100MG
Encompass_Preferred
GNE-781 (compound 19) is an orally active highly potent and selective bromodomain inhibitor of cyclic adenosine monophosphate response element binding protein (CBP) with IC50 of 0 94 nM in TR-FRET assay GNE-781 also inhibits BRET and BRD4(1) with IC50 of 6 2 nM and 5100 nM respectively GNE-781 exhibits antitumor activity
Catalog No. 50-000-01277