Learn More
Medchemexpress LLC GNE-8505 1mg | 1620573-48-9 | 1 MG

Supplier: Medchemexpress LLC HY1143321MG
GNE-8505 is an orally available small-molecule inhibitor of dual leucine zipper kinase (DLK, MAP3K12) used in preclinical research to inhibit DLK signaling and downstream JNK phosphorylation. Supplied as a high-purity, white to off-white solid for laboratory research use only.
- Orally active dual leucine zipper kinase inhibitor
- High purity: 99.86%
- Molecular formula C21H24F3N5O, molecular weight 419.44 g/mol
- White to off-white solid powder
- Storage: powder -20°C (3 years), 4°C (2 years)
- For research use only, not for human or veterinary use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.