Learn More
Medchemexpress LLC GNE-8505 25mg | 1620573-48-9 | 25 MG

Supplier: Medchemexpress LLC HY11433225MG
GNE-8505 is an orally available inhibitor of dual leucine zipper kinase (DLK) supplied as a high-purity research chemical for biochemical and cellular studies targeting DLK/MAP3K signaling.
- High purity: 99.86%.
- Molecular weight: 419.44 g/mol.
- Chemical formula: C21H24F3N5O.
- Available as solid (1 mg, 5 mg, 10 mg, 25 mg) and as a 10 mM solution in DMSO.
- Soluble in DMSO for solution formats.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.