Learn More
Selleck Chemical LLC GNE-9815-E1079-25MG
SDP
Supplier:  Selleck Chemical LLC E107925MG
Encompass_Preferred
GNE-9815 is a highly selective pan-Raf inhibitor with good oral bioavailability GNE-9815 exhibits Ki values of 0 062 and 0 19 nM for c-Raf and b-Raf respectively
Catalog No. 50-313-8528