Learn More
Medchemexpress LLC GNF4877 | 2041073-22-5 | 99.8% | 50 MG

Supplier: Medchemexpress LLC HY12949250MG
GNF4877 is a potent inhibitor of DYRK1A and GSK3β, with IC50 values of 6 nM and 16 nM, respectively. This inhibitory action blocks the nuclear export of nuclear factor of activated T-cells (NFATc) and increases β-cell proliferation, making it suitable for research into metabolic and neurological diseases.
- Potent inhibitor of DYRK1A (IC50 6 nM) and GSK3β (IC50 16 nM)
- Blocks nuclear export of nuclear factor of activated T-cells (NFATc)
- Increases β-cell proliferation (EC50 0.66 μM for mouse β (R7T1) cells)
- Induces β-cell proliferation, increases β-cell mass and insulin content, and improves glycemic control in vivo
- Purity of 99.82%
- Molecular weight of 494.52
- Chemical formula C25H27FN6O4
- Appears as a light yellow to yellow solid
- Soluble in DMSO at 3.33 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.