Learn More
Medchemexpress LLC Gnf6702 | 1799329-72-8 | 98.3% | 50 MG

Supplier: Medchemexpress LLC HY12006050MG
Gnf6702 is a selective inhibitor of the kinetoplastid proteasome that clears parasites in murine models of leishmaniasis, Chagas disease, and human African trypanosomiasis. Supplied as a 50 mg research-use powder, it has a molecular formula of C22H16FN7O2 and a molecular weight of 429.41 g/mol.
- Selective inhibitor of the kinetoplastid proteasome.
- Demonstrated efficacy in murine models of leishmaniasis, Chagas disease, and human African trypanosomiasis.
- Purity reported as 98.33%.
- Soluble in DMSO at 10 mg/mL; in vivo solubility ≥ 1 mg/mL in recommended vehicle.
- Storage: powder stable at -20°C (up to 3 years) and 4°C (up to 2 years); in solution store at -80°C for extended stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.