Learn More
Cayman Chemical GnglIosID GD3bovIn braIn 500ug
SDP
Supplier:  Cayman Chemical 17481500 UG
Encompass_Preferred
A mixture of ganglioside GD3 with C18:1 and C20:1 sphingoid backbones; induces apoptosis in HuT-78 cutaneous T cell lymphoma cells in a concentration-dependent manner and disrupts the mitochondrial membrane potential at 200 μM; expression in GD3-negative SK-MEL-28-N1 malignant melanoma cells increases both cell proliferation and invasion in vitro; ganglioside GD3-deficient adult mice exhibit progressive loss of the NSC pool and impaired neurogenesis
Catalog No. 50-296-7167