Learn More
Cayman Chemical Ganglioside GM1 Monoclonal Antibody (Clone DG2) 100ug
SDP
Supplier:  Cayman Chemical 38290100 UG
Encompass_Preferred
Immunogen: Ganglioside GM1 liposomes • Host: Mouse • Species Reactivity: (+) Species independent • Cross Reactivity: (+) Ganglioside GM1; (-) Other gangliosides • Applications: Dot blot, ELISA, IF, TLC
Catalog No. 50-291-8959