Learn More
Cayman Chemical GnglIosIde GD1bbovInam 5mg
SDP
Supplier:  Cayman Chemical 317085 MG
Encompass_Preferred
A mixture of ganglioside GD1b molecular species isolated from bovine brain with primarily C18:0 fatty acyl chain lengths; a component of plasma membranes; packs densely with cholesterol to form lipid microdomains that modulate both intra- and intercellular signaling events; the concentration of ganglioside GD1b in human brain increases with age
Catalog No. 50-299-2589