Learn More
Cayman Chemical GnglIosIde GQ1bbovInam 100ug
SDP
Supplier:  Cayman Chemical 31561100 UG
Encompass_Preferred
A mixture of ganglioside GQ1b with variable sphingoid backbone chain lengths; stimulates phosphorylation of several ecto-type protein kinase substrates on the surface of GOTO human neuroblastoma cells at 5 nM; promotes differentiation of mESCs to neuronal precursor and glial cells via activation of the ERK1/2 pathway; induces differentiation of murine keratinocytes through phosphoinositide turnover
Catalog No. 50-299-0960