Learn More
Andwin Scientific GNMAT 1.7ML CENT TUBE 10PKG/CS
SDP
Supplier:  Andwin Scientific C32621CS
Encompass_Preferred
The GeneMate 1.7 mL Graduated Microcentrifuge Tubes are ideal for boiling applications. The tubes work within the temperature range of -90C to 120C. This product features wide cap flange that permits easy opening. The tubes are superior in clarity and are free of detectable DNase, RNase, and human DNA. It is sold in clear color.
Catalog No. NC0102719