Learn More
Cayman Chemical GnoderenIc AcId A 10mg
SDP
Supplier:  Cayman Chemical 2768510 MG
Encompass_Preferred
A triterpenoid; an inhibitor of β-glucuronidase (IC50 = 0.12 mg/ml in rat liver microsomes); reduces increases in serum AST, ALT, and LDH in a CCl4-induced rat model of liver injury at 100 mg/kg; inhibits aldose reductase by 54.2% at 100 UG/ml
Catalog No. 50-294-0185