Learn More
Cayman Chemical GnoderIc AcId A 10mg
SDP
Supplier:  Cayman Chemical 2667410 MG
Encompass_Preferred
A triterpene with diverse biological activities; reduces cell viability, expression SOD1, SOD2, and SOD3, and production of ROS in PC3 prostate cancer cells; inhibits LPS-induced activation of NF-κB, release of TNF-α, IL-1β, and IL-6, and mitochondrial activity in primary mouse cortical microglia; reduces lung MPO activity, neutrophil infiltration, and NF-κB signaling, as well as BALF levels of TNF-α, IL-1β, and IL-6 in a mouse model of LPS-induced lung injury at 20 and 40 mg/kg; reduces weight gain and hepatic lipid accumulation and improves insulin sensitivity in a mouse model of high-fat diet-induced obesity
Catalog No. 50-293-4175