Learn More
Cayman Chemical GnoderIc AcId H 5mg
SDP
Supplier:  Cayman Chemical 297035 MG
Encompass_Preferred
A triterpene with diverse biological activities; inhibits ACE (IC50 = 26 µM); inhibits HIV-1 protease (IC50 = 200 µM); reduces colony formation and migration of MDA-MB-231 breast cancer cells; inhibits acetic acid-induced writhing in mice at 1, 3, and 5 mg/kg
Catalog No. 50-298-8996