Learn More
Apexbio Technology LLC GnRH Associated Peptide (GAP) (1-13), human 100111-07-7 10mg
SDP
Supplier:  Apexbio Technology LLC A102010
Encompass_Preferred
GnRH Associated Peptide (GAP) (1-13) human (CAS 100111-07-7) corresponds to the amino acid sequence Asp-Ala-Glu-Asn-Leu-Ile-Asp-Ser-Phe-Gln-Glu-Ile-Val derived from the N-terminal region of human GAP This peptide originates from the precursor of gonadotropin-releasing hormone (GnRH) which is co-released with GnRH from hypothalamic neurons into the pituitary portal system GAP (1-13) has been shown to suppress prolactin secretion and stimulate gonadotropin release implicating a modulatory role in neuroendocrine pathways governing reproductive and pituitary function The peptide is utilized in research on hypothalamic-pituitary axis regulation neuroendocrine signaling and mechanisms controlling hormone secretion
Catalog No. 50-000-93298