Learn More
Selleck Chemical LLC Go 6983 10mg 133053-19-7 GOE 6983
SDP
Supplier:  Selleck Chemical LLC S291110MG
Encompass_Preferred
Go 6983 (GOE 6983) is a pan-PKC inhibitor against for PKC?, PKC?, PKC? and PKC? with IC50 of 7 nM, 7 nM, 6 nM and 10 nM, respectively; less potent to PKC? and inactive to PKC?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3190