Learn More
Medchemexpress LLC HY-13689 10mg , Go 6983 CAS:133053-19-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1368910MG
Encompass_Preferred
HY-13689 10mg Go 6983 CAS: 133053-19-7 Go 6983 is a pan-PKC inhibitor against for PKCα, PKCβ, PKCγ, PKCδ and PKCζ with IC50 of 7 nM, 7 nM, 6 nM, 10 nM and 60 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9716