Learn More
Selleck Chemical LLC Go6976 5mg 136194-77-9 PD406976
SDP
Supplier:  Selleck Chemical LLC S71195MG
Encompass_Preferred
Go6976 (PD406976) is a potent PKC inhibitor with IC50 of 7.9 nM, 2.3 nM, and 6.2 nM for PKC (Rat brain), PKC?, and PKC?1, respectively. Also a potent inhibitor of JAK2 and Flt3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4424