Learn More
Selleck Chemical LLC Go6976
SDP
Supplier:  Selleck Chemical LLC S71191G
Encompass_Preferred
Go6976 (PD406976) is a potent PKC inhibitor with IC50 of 7 9 nM 2 3 nM and 6 2 nM for PKC (Rat brain) PKC and PKC 1 respectively Also a potent inhibitor of JAK2 and Flt3
Catalog No. 50-000-02229