Learn More
Immunoreagents Inc GOAT A-CHICKEN IGG BIO 2 MG
Supplier:  Immunoreagents Inc GTXCK003DBIO
Encompass_Preferred
Secondary Antibody; Goat anti-chicken IgG/Y (H&L), Affinity Pure, Biotin Conjugate, Host: Goat, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-089