Learn More
Immunoreagents Inc GOAT A-DONKEY IGG DYLIGHT 633
Supplier:  Immunoreagents Inc GTXDK003D633NHSX
Encompass_Preferred
Secondary Antibody; Goat anti-donkey IgG (H&L), Affinity Pure, DyLight 633 Conjugate, Host: Goat, Format: Lyo, Label: DyL633, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-270-900