Learn More
Immunoreagents Inc GOAT A-HUMAN IG KAPPA ALP
Supplier:  Immunoreagents Inc GTXHU084DALP
Encompass_Preferred
Secondary Antibody; Goat anti-human (kappa light chain) - Affinity Pure, ALP Conjugate, Host: Goat, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-515