Learn More
Immunoreagents Inc GOAT A-HUMAN IGG ALP 0.5 MG
Supplier:  Immunoreagents Inc GTXHU004JALP
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, Fc, F(ab)'2 fragment - Affinity Pure, min x w/Bo, Eq, Mu or Rb serum proteins, ALP Conjugate, Host: Goat, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-921