Learn More
Immunoreagents Inc GOAT A-HUMAN IGG ALP 1 MG
Supplier:  Immunoreagents Inc GTXHU004FALP
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, Fc - Affinity Pure, min x w/human IgA + IgM, ALP Conjugate, Host: Goat, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-477