Learn More
Immunoreagents Inc GOAT A-HUMAN IGG AP 2 MG
Supplier:  Immunoreagents Inc GTXHU003E3
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, (H&L), Affinity Pure, min x w/rat IgG, Host: Goat, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-263