Learn More
Immunoreagents Inc GOAT A-HUMAN IGG HRP 0.5 MG
Supplier:  Immunoreagents Inc GTXHU003GHRPX
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, (H&L), F(ab)'2 fragment - Affinity Pure, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-270-964