Learn More
Immunoreagents Inc GOAT A-HUMAN IGG HRP 1 MG
Supplier:  Immunoreagents Inc GTXHU003E2HRPX
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, (H&L), Affinity Pure, min x w/Bo, Gt, Mu or Rb serum proteins, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-270-949