Learn More
Immunoreagents Inc GOAT A-HUMAN IGM MU AP
Supplier:  Immunoreagents Inc GTXHU006E2
Encompass_Preferred
Secondary Antibody; Goat anti-human IgM, (muchain), Affinity Pure, min x w/Human IgG; Bo, Gt, Mu or Rb Serum Proteins, Host: Goat, Format: Liq, Label: None , Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-270-979