Learn More
Immunoreagents Inc GOAT A-MOUSE IGG DYLIGHT 350
Supplier:  Immunoreagents Inc GTXMU003E2350NHSX
Encompass_Preferred
Secondary Antibody; Goat anti-mouse IgG (H&L) - Affinity Pure, min x w/Bo, Eq, Hu, Pig or Rb Serum Proteins, DyLight 350 Conjugate, Host: Goat, Format: Lyo, Label: DyL350, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-272-147