Learn More
Immunoreagents Inc GOAT A-MOUSE IGM HRP
Supplier:  Immunoreagents Inc GTXMU007DHRPX
Encompass_Preferred
Secondary Antibody; Goat anti-mouse IgM (H&L) - Affinity Pure, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/IC
Catalog No. 50-271-275