Learn More
Immunoreagents Inc GOAT A-RAT IGG FITC
Supplier:  Immunoreagents Inc GTXRT003JFITC
Encompass_Preferred
Secondary Antibody: Goat anti-rat IgG (H&L), F(ab)'2 fragment - Affinity Pure, min x w/Hu or Mu IgG, FITC Conjugate, Host: Goat, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-953