Learn More
Immunology Consultants Affinity Purified Goat anti-Cat IgG Fc Antibody, 1.0 mg
SDP
Supplier:  Immunology Consultants GGFC20A
Encompass_Preferred
Goats were immunized with highly purified cat IgG Fc fragment and the resulting antiserum was collected. Antibody was solid phase adsorbed and immunoaffinity purified off a catIgG containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with cat IgG and not cat IgG Fab fragment as determined by IEP and ELISA techniques.It is suitable for blotting, ELISA, and IHC applications. This antibody may cross-react with IgG from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-224-8889