Learn More
Immunology Consultants Affinity Purified Goat anti-Human IgE Antibody, 1.0 mg
SDP
Supplier:  Immunology Consultants GE80A
Encompass_Preferred
Goats were immunized with highly purified human IgE and antiserum was collected. Antibody was solid phase adsorbed and immunoaffinity purified off a human IgE containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with humanIgE as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA, and IHC applications. This antibody may cross-react with IgE from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2182