Learn More
Medchemexpress LLC Goat Anti-Mouse IgG 100uL | 100uL
SDP
Supplier:  Medchemexpress LLC HYP87169100UL
Encompass_Preferred
Goat Anti-Mouse IgG H L is a non-conjugated and Goat origined monoclonal antibody targeting to Mouse IgG antibody Goat Anti-Mouse IgG H L can binds to the light and heavy chains of Mouse IgG antibodies
Catalog No. 50-003-25339