Learn More
Spherotech GOAT ANTI-MOUSE IGG (H&L) COAT
Supplier:  Spherotech MP605
Encompass_Preferred
Goat anti-Mouse IgG (H&L) Coated Polystyrene Particles. 0.5% w/v.6.0-8.0 µm.5 mL
Catalog No. NC1784783