Learn More
Bioworld GOAT ANTI-MOUSE IGG SEPARO
Supplier:  Bioworld 201810911
Encompass
GOAT ANTI-MOUSE IGG SEPAROPORE (AGAROSE) 4B: Affinity isolated antibody is coupled to cyanogen bromide-activated Separopore4B. The antibody preparation is specific for the Fc fragment of mouse IgG when tested against purified mouse IgA,...
Catalog No. 50-197-901