Learn More
Abcam Goat Anti-Rabbit IgG H&L (Alexa Fluor 488)
Supplier:  Abcam AB150077
Encompass_Preferred
Goat anti-rabbit IgG H&L (Alexa Fluor 488) is a secondary antibody with a maximum absorption wavelength of 495nm and a maximum emission wavelength of 519nm. Ideal for fluorescent cell and tissue imaging. Suitable for ICC/IF, IHC, flow cytometry and ELISA.- Intense fluorescence and high photostability allowing more time for image capture especially when detecting low abundance targets- Minimal spectral overlap with other Alexa Fluor dyes makes this ideal for use in multi-color analysis- Cited in over 3100 publications
Catalog No. NC3906351