Learn More
Abcam Goat Anti-Rabbit IgG H&L (HRP)
Supplier:  Abcam AB6721
Encompass_Preferred
Goat anti-rabbit IgG H&L (HRP) (ab6721) is a polyclonal horseradish peroxidase (HRP)-conjugated secondary antibody. Suitable for western blot, ICC, IHC, ELISA and Dot blot applications.- Flexible signal amplification- use in colorimetric or chemiluminescent detection- Proven performance: cited in over 5940 publications
Catalog No. NC3871866