Learn More
GenDEPOT Goat anti-Rat IgG
Supplier:  GenDEPOT SA006500
Encompass_Preferred
Affinity purified antibody isolated from a pool of serum from goats immunizedwith purified rat IgG was labeled with peroxidase by the periodate method of Nakaneand Kawaoi
Catalog No. 50-190-4210