Learn More
Rockland Immunochemicals Anti-RAT IgG (H&L) (GOAT) Antibody DyLight™ 800 Conjugated - 612-145-002
SDP
Supplier:  Rockland Immunochemicals 612145002
Encompass_Preferred
Anti-Rat IgG (H&L) DyLight™800 Antibody generated in goat detects reactivity to Rat IgG. This antibody will react with heavy chains of Rat IgG and with light chains of most Rat immunoglobulins. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This Anti-Rat IgG is conjugated to DyLight™800. This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
Catalog No. NC0977875