Learn More
Jackson Immuno Research Labs Biotin-SP (long spacer) AffiniPure Goat Anti-Rat IgG (H+L), .5ml
SDP
Supplier:  Jackson Immuno Research Labs 112065167
Encompass_Preferred
Biotin-SP-AffiniPure Goat Anti-Rat IgG (H+L) (min X Ms,Hu,Bov,Hrs,Rb Sr Prot)
Catalog No. NC9177931