Learn More
Vector Laboratories Goat Anti-Wheat Germ Agglutinin (WGA), Unconjugated, 1 mg
SDP
Supplier:  Vector Laboratories AS20241
Encompass
Goat Anti-Wheat Germ Agglutinin (WGA), Unconjugated, 1 mg, antibodies to lectins are produced by immunizing goats with pure lectins. Following conventional purification steps, specific antibody is isolated by affinity chromatography on lectin-agarose columns. This anti-lectin is supplied in solution. These highly purified specific antibodies provide excellent intermediate reagents for localizing lectin receptors in tissues and on cells or following lectin transport in neuronal tracing.
Catalog No. NC1856721