Learn More
Cayman Chemical Goat AntiRabbit IgG FITC
SDP
Supplier:  Cayman Chemical 100065881 EA
Encompass_Preferred
Immunogen: Mouse IgG • Host: Goat • Application: IF • This goat anti-rabbit IgG FITC is used as the 'secondary antibody' for immunofluorescent staining on tissues or culture cells where the primary antibody was generated in rabbits. It is highly recommended that proper negative controls, such as omitting the primary antibody, be included in experiments when conducting immunofluorescent staining.
Catalog No. 50-288-7051