Learn More
Cayman Chemical Goat AntiRabbit IgG HRP
SDP
Supplier:  Cayman Chemical 100043011 EA
Encompass_Preferred
This goat anti-rabbit IgG HRP is used as the 'secondary antibody' for western blotting or ELISA where the primary antibody was generated in rabbits. This peroxidase-conjugated secondary antibody is suitable for western blotting when employing detection by commercially available chemiluminescence substrates. Additionally this secondary antibody conjugate can be used to develop ELISAs with substrates like ABTS (2,2'-azino-bis-[3-ethylbenzothiazoline-6-sulfonic acid]-diammonium salt), OPD (O-phenylenediamine), or TMB (3,3,5,5-tetramethylbenzidine).
Catalog No. 50-288-6519