Learn More
Cayman Chemical Goat F Ab2 ntI-Mouse IgG 100ug
SDP
Supplier:  Cayman Chemical 16647100 UG
Encompass_Preferred
Goat F(Ab')2 anti-Mouse IgG, H+L conjugated to SureLight™ R-PE Antibody: Goat F(Ab')2 polyclonal anti-Mouse IgG Isotype: IgG Dye: SureLight™ R-phycoerythrin Excitation max. λ: 565>498 nm Emission max. λ: 578 nm Uses: Flow cytometry, Fluorescence-activated cell sorting, Luminex, and Protein Microarrays
Catalog No. 50-296-8196