Learn More
Vector Laboratories Goat Anti-PHA E+L Unconjugated I-5000-5
SDP
Supplier:  Vector Laboratories I50005
Encompass
Goat Anti-Phaseolus Vulgaris Agglutinin (E+L), Unconjugated, 1 mg, antibodies to lectins are produced by immunizing goats with pure lectins. Following conventional purification steps, specific antibody is isolated by affinity chromatography on lectin-agarose columns. Each anti-lectin is supplied lyophilized in buffered saline. These highly purified specific antibodies provide excellent intermediate reagents for localizing lectin receptors in tissues and on cells or following lectin transport in neuronal tracing.
Catalog No. 50-204-5663