Learn More
Cayman Chemical Goat ntI-Humn IgGDyLIght 1mg
SDP
Supplier:  Cayman Chemical 166991 MG
Encompass_Preferred
Goat anti-human IgG, secondary antibody, conjugated to DyLight 488Antibody: Goat polyclonal anti-Human IgG Dye: DyLight 488 Excitation max. λ: 493 nm Emission max. λ: 518 nm Uses: Flow Cytometry, flurorescence-activated cell sorting, and protein microarrays Form: Conjugate supplied as a liquid in 0.1 M sodium phosphate, 0.15 M NaCl (pH 7.4), 1% BSA, and 0.02% NaN3.
Catalog No. 50-296-8199